Sinapic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid
A15676.03
A15676.06
A15676.14
A15676.03EA
1120
CZK
A15676.03
A15676.06
A15676.14
Sinapic acid ≥98%
Sinapic acid
Purity: 98%
CAS No.: 530-59-6
Molecular Formula: C11H12O5
Formula Weight: 224.21
Melting Point: ca 195° dec.
Beilstein Reference: 2130006
Vzorec:
C₁₁H₁₂O₅ MW: 224,21 g/mol Bod varu: 378 °C (1013 hPa) Bod tání: ∼ 195 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00004401 CAS číslo: 530-59-6 EINECS: 208-487-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...