Tisk…

trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

M-Clarity™ quality level = MQ200

128708-100G 128708-25G 128708-5G
SIAL128708-100GEA 5020 CZK
SIAL128708-100G SIAL128708-25G SIAL128708-5G
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, Sigma-Aldrich®
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid
Vzorec: C₁₀H₁₀O₄
MW: 194,18 g/mol
MDL Number: MFCD00004400
CAS číslo: 537-98-4
EINECS: 208-679-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR