4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), tan powder
Dodavatel: MP Biomedicals
Synonyms:
3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid
Ferulic acid is a chemopreventive caffeic acid analogue. Ferulic acid is an inhibitor of 5-LO (5-lipoxygenase) and 12-LO (12-lipoxygenase).
Ferulic acid is an antioxidant which neutralises free radicals (superoxide, nitric oxide and hydroxyl radical) which could cause oxidative damage of cell membranes and DNA.
Soluble in hot water. (Lit.)
Vzorec:
C₁₀H₁₀O₄ MW: 194,18 g/mol Bod tání: 168…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004400 CAS číslo: 1135-24-6 EINECS: 208-679-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...