Tisk…

trans-4-Methoxycinnamic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: trans-O-Methyl-p-coumaric Acid, (E)-3-(4-Methoxyphenyl)-2-propenoic acid

A13870.18 A13870.09 A13870.30
A13870.18EA 2540 CZK
A13870.18 A13870.09 A13870.30
trans-4-Methoxycinnamic acid 98%
trans-4-Methoxycinnamic acid

Purity: 98% CAS No.: 943-89-5 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 171-175° Beilstein Reference: 510468

Vzorec: C₁₀H₁₀O₃
MW: 178,19 g/mol
Bod varu: 317 °C (1013 hPa)
Bod tání: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004398
CAS číslo: 830-09-1
EINECS: 213-405-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR