Tisk…

3-Methoxycinnamic acid predominantly trans ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(3-Methoxyphenyl)acrylic acid

A10723.06 A10723.14 A10723.22
A10723.06EA 841 CZK
A10723.06 A10723.14 A10723.22
3-Methoxycinnamic acid predominantly trans ≥98%
3-Methoxycinnamic acid

Purity: 98+% CAS No.: 6099-04-3 Molecular Formula: C10H10O3 Formula Weight: 178.19 Melting Point: 117-121° Beilstein Reference: 2209732

Vzorec: C₁₀H₁₀O₃
MW: 178,19 g/mol
Bod tání: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00004386
CAS číslo: 6099-04-3
EINECS: 228-049-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR