Tisk…

trans-Cinnamic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A13538.0E A13538.22 A13538.36
A13538.0EEA 10710 CZK
A13538.0E A13538.22 A13538.36
trans-Cinnamic acid ≥99%
trans-Cinnamic acid

Purity: 99+% CAS No.: 140-10-3 Molecular Formula: C9H8O2 Formula Weight: 148.16 Flash Point: 166°(330°F) Physical Form: Powder Density: 1.250 Appearance: White to pale cream Odour: Weak, characteristic Melting Point: 133-136° Boiling Point: 147°/4mm Beilstein Reference: 1905952 Merck Reference: 14,2299 MDL No.: MFCD00004369

Vzorec: C₉H₈O₂
Bod varu: 147 °C (4 mmHg)
Bod tání: 133…136 °C
Hustota: 1,250
Bod vzplanutí: 166 °C (330 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004369
CAS číslo: 140-10-3
EINECS: 205-398-1
Merck Index: 14,2299

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR