4-Chlorocinnamic acid predominantly trans 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(4-Chlorophenyl)acrylic acid
A15304.14
A15304.22
A15304.14EA
2220
CZK
A15304.14
A15304.22
4-Chlorocinnamic acid predominantly trans 99%
4-Chlorocinnamic acid
Purity: 99%
CAS No.: 1615-02-7
Molecular Formula: C9H7ClO2
Formula Weight: 182.61
Melting Point: 248-253°
Beilstein Reference: 1365308
Vzorec:
C₉H₇ClO₂ MW: 182,61 g/mol Bod varu: 311 °C (1013 hPa) Bod tání: 248…253 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004396 CAS číslo: 1615-02-7 EINECS: 216-564-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...