Tisk…

4-Bromocinnamic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 3-(4-Bromophenyl)acrylic acid

M-Clarity™ quality level = MQ100

260975-25G 260975-5G
SIAL260975-25GEA 4380 CZK
SIAL260975-25G SIAL260975-5G
4-Bromocinnamic acid, Sigma-Aldrich®
4-Bromocinnamic acid
Vzorec: C₉H₇BrO₂
MW: 227,06 g/mol
MDL Number: MFCD00004394
CAS číslo: 1200-07-3
EINECS: 214-850-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR