4-Bromocinnamic acid predominantly trans ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(4-Bromophenyl)acrylic acid
A11503.06
A11503.14
A11503.22
A11503.06EA
1160
CZK
A11503.06
A11503.14
A11503.22
4-Bromocinnamic acid predominantly trans ≥98%
4-Bromocinnamic acid
Purity: 98+%
CAS No.: 1200-07-3
Molecular Formula: C9H7BrO2
Formula Weight: 227.06
Odour: Odourless
Melting Point: 260-264°
Beilstein Reference: 3538823
MDL No.: MFCD00004394
Vzorec:
C₉H₇BrO₂ MW: 227,06 g/mol Bod tání: 260…264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004394 CAS číslo: 1200-07-3 EINECS: 214-850-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...