2-Methyl-1-butene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98%
CAS No.: 563-46-2
Molecular Formula: C5H10
Formula Weight: 70.13
Storage Temperature: 2°C to 8°C
Flash Point: -48°(-54°F)
Refractive Index: 1.3740
Melting Point: -137°
Boiling Point: 31-32°
UN No.: 2459
MDL No.: MFCD00009333
Vzorec:
C₅H₁₀ Bod varu: 31…32 °C Bod tání: –137 °C Hustota: 0,651 Bod vzplanutí: –48 °C (–54 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009333 CAS číslo: 563-46-2 EINECS: 209-250-7 UN: 2459 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...