Tisk…

trans-4-Octene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

593028-25G 593028-5G
SIAL593028-25GEA 13910 CZK
SIAL593028-25G SIAL593028-5G
trans-4-Octene, Sigma-Aldrich®
trans-4-Octene
Vzorec: C₈H₁₆
Bod varu: 122 °C (1013 hPa)
Hustota: 0,7141 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009476
CAS číslo: 14850-23-8
EINECS: 238-913-3
UN: 3295
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR