Tisk…

cis-2-Pentene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

143766-10G 143766-1G
SIAL143766-10GEA 7050 CZK
SIAL143766-10G SIAL143766-1G
cis-2-Pentene, Sigma-Aldrich®
cis-2-Pentene
Vzorec: C₅H₁₀
Bod varu: 37 °C (1013 hPa)
Hustota: 0,6556 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063978
CAS číslo: 627-20-3
EINECS: 210-988-7
UN: 3295
ADR: 3,II
Merck Index: 12,07263

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR