1-Octadecene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
O806-1L
O806-25ML
SIALO806-1LEA
2250
CZK
SIALO806-1L
SIALO806-25ML
1-Octadecene, Sigma-Aldrich®
1-Octadecene
Vzorec:
C₁₈H₃₆ Bod varu: 315 °C (1013 hPa) Hustota: 0,789 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009003 CAS číslo: 112-88-9 EINECS: 204-012-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...