Tisk…

trans-4-Hexen-1-ol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (E)-Hex-4-en-1-ol, trans-Hex-4-en-1-ol

M-Clarity™ quality level = MQ100

237604-25G 237604-5G
SIAL237604-25GEA 24580 CZK
SIAL237604-25G SIAL237604-5G
trans-4-Hexen-1-ol, Sigma-Aldrich®
trans-4-Hexen-1-ol
Vzorec: C₆H₁₂O
MW: 100,16 g/mol
Bod varu: 157…159 °C (1013 hPa)
Hustota: 0,851 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009713
CAS číslo: 928-92-7
EINECS: 213-188-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR