trans-3-Hexen-1-ol ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
L10541.06
L10541.14
L10541.06EA
2060
CZK
L10541.06
L10541.14
trans-3-Hexen-1-ol ≥97%
trans-3-Hexen-1-ol
Purity: 97%
CAS No.: 928-97-2
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 58°(136°F)
Refractive Index: 1.4390
Odour: Pleasant
Boiling Point: 60-61°/12mm
Beilstein Reference: 1719713
Merck Reference: 14,4700
UN No.: 1987
MDL No.: MFCD00002960
Vzorec:
C₆H₁₂O Bod varu: 60…61 °C (12 mmHg) Bod tání: –60 °C Hustota: 0,845 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002960 CAS číslo: 928-97-2 EINECS: 213-193-3 UN: 1987 ADR: 3,III Merck Index: 14,4700 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...