trans-2-Octen-1-ol ≥97%
Dodavatel: Thermo Fisher Scientific
Varování
|
Synonyms:
Trans-2-octen-1-ol, trans-2-Octen-1-ol, trans-Oct-2-en-1-ol
A15960.06
A15960.14
A15960.22
A15960.06EA
1630
CZK
A15960.06
A15960.14
A15960.22
trans-2-Octen-1-ol ≥97%
trans-2-Octen-1-ol
Purity: 97%
CAS No.: 18409-17-1
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 85°(185°F)
Refractive Index: 1.4465
Boiling Point: 88°/11mm
Beilstein Reference: 1720707
MDL No.: MFCD00014057
Vzorec:
C₈H₁₆O MW: 128.21 Bod varu: 88 °C (11 mmHg) Hustota: 0.850 Bod vzplanutí: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014057 CAS číslo: 18409-17-1 EINECS: 242-288-2 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...