trans-2-Hexen-1-ol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
trans-Hex-2-en-1-ol
A13272.14
A13272.22
A13272.36
A13272.14EA
1410
CZK
A13272.14
A13272.22
A13272.36
trans-2-Hexen-1-ol ≥97%
trans-2-Hexen-1-ol
Purity: 97%
CAS No.: 928-95-0
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 54°(129°F)
Refractive Index: 1.4380
Odour: Pleasant
Boiling Point: 158-160°
Beilstein Reference: 1719709
UN No.: 1987
MDL No.: MFCD00002927
Vzorec:
C₆H₁₂O MW: 100.16 Bod varu: 158…160 °C Bod tání: 65…67 °C Hustota: 0.843 Bod vzplanutí: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002927 CAS číslo: 928-95-0 EINECS: 213-191-2 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...