Tisk…

4-Penten-1-ol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A11178.14 A11178.22 A11178.06 AB118419.0025
A11178.14EA 6040 CZK
A11178.14 A11178.22 A11178.06 ABCRAB118419.0025
4-Penten-1-ol ≥98%
4-Penten-1-ol

Purity: 98+% CAS No.: 821-09-0 Molecular Formula: H2C=CH(CH2)3OH Formula Weight: 86.13 Flash Point: 43°(109°F) Physical Form: Liquid Density: 0.840 Refractive Index: 1.4300 Boiling Point: 134-137° Beilstein Reference: 1560163 UN No.: 1987 MDL No.: MFCD00002975

Vzorec: C₅H₁₀O
Bod varu: 134…137 °C
Hustota: 0,840
Bod vzplanutí: 43 °C (109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002975
CAS číslo: 821-09-0
EINECS: 212-473-2
UN: 1987
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR