Tisk…

3-Buten-1-ol, Sigma-Aldrich®

Dodavatel: Merck

496839-100G 496839-1KG 496839-25G
SIAL496839-100GEA 9610 CZK
SIAL496839-100G SIAL496839-1KG SIAL496839-25G
3-Buten-1-ol, Sigma-Aldrich®
3-Buten-1-ol
Vzorec: C₄H₈O
MDL Number: MFCD00002959
CAS číslo: 627-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR