Tisk…

3-Buten-1-ol ≥98%

Dodavatel: Thermo Fisher Scientific

L00767.06 L00767.14
L00767.06EA 1660 CZK
L00767.06 L00767.14
3-Buten-1-ol ≥98%
3-Buten-1-ol

Purity: 98+% CAS No.: 627-27-0 Molecular Formula: CH2=CHCH2CH2OH Formula Weight: 72.11 Flash Point: 32°(89°F) Refractive Index: 1.4210 Boiling Point: 113-114° Beilstein Reference: 1633504 UN No.: 1987 MDL No.: MFCD00002959

Vzorec: C₄H₈O
MDL Number: MFCD00002959
CAS číslo: 627-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR