Tisk…

3-Methoxypropene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Allyl methyl ether

M-Clarity™ quality level = MQ100

678201-25G 678201-5G
SIAL678201-25GEA 13180 CZK
SIAL678201-25G SIAL678201-5G
3-Methoxypropene, Sigma-Aldrich®
3-Methoxypropene
Vzorec: C₄H₈O
MW: 72,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008649
CAS číslo: 627-40-7
EINECS: 210-997-6
UN: 3271
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR