Tisk…

Mesaconic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

131040-10G 131040-50G
SIAL131040-10GEA 3550 CZK
SIAL131040-10G SIAL131040-50G
Mesaconic acid, Sigma-Aldrich®
Mesaconic acid
Vzorec: C₅H₆O₄
Bod tání: 200 °C
CAS číslo: 498-24-8
EINECS: 207-859-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR