Citraconic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Methyl-2-butenedioic acid
L04178.09
L04178.18
L04178.09EA
1100
CZK
L04178.09
L04178.18
Citraconic acid ≥98%
Citraconic acid
Purity: 98+%
CAS No.: 498-23-7
Molecular Formula: C5H6O4
Formula Weight: 130.10
Melting Point: 88-91°
Beilstein Reference: 1722679
Merck Reference: 14,2321
MDL No.: MFCD00078086
Vzorec:
C₅H₆O₄ MW: 130,1 g/mol Bod varu: 336 °C (1013 hPa) Bod tání: 88…91 °C Hustota: 1,620 Storage Temperature: Ambient |
MDL Number:
MFCD00078086 CAS číslo: 498-23-7 EINECS: 207-858-7 Merck Index: 14,2321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...