Citraconic acid 99+%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Methyl-2-butenedioic acid
110430250
110431000
110430050
ACRO110430250EA
1980
CZK
ACRO110430250
ACRO110431000
ACRO110430050
Citraconic acid 99+%
Citraconic acid
Vzorec:
C₅H₆O₄ MW: 130.1 Bod varu: 336 Bod tání: 88…92 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00078086 CAS číslo: 498-23-7 EINECS: 207-858-7 Merck Index: 13,02345 |
Výsledky specifikačního testu
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥99.0 % |
UV Lambda max | 630 to 635 nm (in water) |
Absorption Ratio P-15/P+15 | 0.86 to 1.11 |
Visual transition interval | from pH 0.2 (yellow) to pH 1.8 (blue) |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...