Kyselina olejová ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(9Z)-9-Octadecenoic acid
31997.06
31997.14
31997.22
31997.06EA
2460
CZK
31997.06
31997.14
31997.22
Kyselina olejová ≥99%
Kyselina olejová
Purity: 99%
CAS No.: 112-80-1
Molecular Formula: C18H34O2
Formula Weight: 282.47
Physical Form: Liquid
Density: 0.887
Refractive Index: 1.4610
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Melting Point: 13-14°
Boiling Point: 286°/100mm
Beilstein Reference: 1726542
Merck Reference: 14,6828
MDL No.: MFCD00064242
Vzorec:
C₁₈H₃₄O₂ MW: 282.47 Bod varu: 286 °C (100 mmHg) Bod tání: 13…14 °C Hustota: 0.887 Bod vzplanutí: 180 Storage Temperature: Ambient |
MDL Number:
MFCD00064242 CAS číslo: 112-80-1 EINECS: 204-007-1 Merck Index: 13,06898 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...