4-Pentenoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Allylacetic acid, Pent-4-enoic acid
M-Clarity™ quality level = MQ300
W284300-100G-K
W284300-1KG-K
W284300-25G-K
SIALW284300-100G-KEA
4100
CZK
SIALW284300-100G-K
SIALW284300-1KG-K
SIALW284300-25G-K
4-Pentenoic acid, Sigma-Aldrich®
4-Pentenoic acid
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 187 °C (1013 hPa) Bod tání: –18 °C Hustota: 0,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004408 CAS číslo: 591-80-0 EINECS: 209-732-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...