Tisk…

Allyl acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

185248-100ML 185248-500ML
SIAL185248-100MLEA 1450 CZK
SIAL185248-100ML SIAL185248-500ML
Allyl acetate, Sigma-Aldrich®
Allyl acetate
Vzorec: C₅H₈O₂
MW: 100,12 g/mol
Bod varu: 103 °C (1013 hPa)
Bod tání: –96 °C
Hustota: 0,93 g/cm³ (20 °C)
Bod vzplanutí: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008721
CAS číslo: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3,II
Merck Index: 12,00293

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR