Tisk…

Fumaroyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

151386-100G 151386-25G
SIAL151386-100GEA 4010 CZK
SIAL151386-100G SIAL151386-25G
Fumaroyl chloride, Sigma-Aldrich®
Fumaroyl chloride
Vzorec: C₄H₂Cl₂O₂
Bod varu: 161 °C (1013 hPa)
Bod tání: –2 °C
Hustota: 1,41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000733
CAS číslo: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR