trans-2-Undecenal, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(E)-2-Undecenal
M-Clarity™ quality level = MQ300
W342300-100G-K
W342300-1KG-K
SIALW342300-100G-KEA
6100
CZK
SIALW342300-100G-K
SIALW342300-1KG-K
trans-2-Undecenal, Sigma-Aldrich®
trans-2-Undecenal
Vzorec:
C₁₁H₂₀O MW: 168,28 g/mol |
MDL Number:
MFCD00014680 CAS číslo: 53448-07-0 EINECS: 258-559-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...