trans-2-Heptenal, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W316504-1KG-K
W316504-200G-K
SIALW316504-1KG-KEA
25230
CZK
SIALW316504-1KG-K
SIALW316504-200G-K
trans-2-Heptenal, Sigma-Aldrich®
trans-2-Heptenal
Vzorec:
C₇H₁₂O Bod varu: 90 °C (1013 hPa) Hustota: 0,8575 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007010 CAS číslo: 18829-55-5 EINECS: 242-608-0 UN: 1988 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...