10-Undecenal, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Undecylenic aldehyde, Undec-10-enal, 10-Undecenyl aldehyde, 10-Undecenal
M-Clarity™ quality level = MQ100
132276-100G
132276-25G
SIAL132276-100GEA
0
CZK
SIAL132276-100G
SIAL132276-25G
10-Undecenal, Sigma-Aldrich®
10-Undecenal
Vzorec:
C₁₁H₂₀O MW: 168,28 g/mol Bod varu: 240 °C (1013 hPa) Bod tání: 7 °C Hustota: 0,8467 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007032 CAS číslo: 112-45-8 EINECS: 203-973-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...