5-Bromo-1-pentene, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
253901-25G
253901-5G
SIAL253901-25GEA
5910
CZK
SIAL253901-25G
SIAL253901-5G
5-Bromo-1-pentene, Sigma-Aldrich®
5-Bromo-1-pentene
Vzorec:
C₅H₉Br MW: 149.03 Bod varu: 126 Hustota: 1.2547 Storage Temperature: Refrigerator |
MDL Number:
MFCD00000264 CAS číslo: 1119-51-3 EINECS: 214-281-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...