2-Bromopropene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide
M-Clarity™ quality level = MQ100
B78254-10G
B78254-50G
SIALB78254-10GEA
3030
CZK
SIALB78254-10G
SIALB78254-50G
2-Bromopropene, Sigma-Aldrich®
2-Bromopropene
Vzorec:
C₃H₅Br MW: 120,98 g/mol Bod varu: 48 °C (1013 hPa) Bod tání: –125 °C Hustota: 1,39 g/cm³ (9 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000140 CAS číslo: 557-93-7 EINECS: 209-185-4 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...