Tisk…

HEPES sodium salt, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, HEPES sodium, HEPES Na

M-Clarity™ quality level = MQ400

H7006-100G H7006-1KG H7006-25G H7006-500G H7006-2.5KG
SIALH7006-100GEA 10780 CZK
SIALH7006-100G SIALH7006-1KG SIALH7006-25G SIALH7006-500G SIALH7006-2.5KG
HEPES sodium salt, Sigma-Aldrich®
HEPES sodium salt
Vzorec: C₈H₁₇N₂NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS číslo: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR