(3-Fluorophenyl)acetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(3-Fluorophenyl)acetic acid
A15635.06
A15635.14
A15635.22
A15635.06EA
1710
CZK
A15635.06
A15635.14
A15635.22
(3-Fluorophenyl)acetic acid ≥98%
(3-Fluorophenyl)acetic acid
Purity: 98%
CAS No.: 331-25-9
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point:
Vzorec:
(C₆H₄F)CH₂COOH MW: 154.14 Bod varu: 151 °C (0.8 mmHg) Bod tání: 44…48 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004331 CAS číslo: 331-25-9 EINECS: 206-360-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...