Tisk…

(2,6-Difluorophenyl)acetic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,6-Difluorophenylacetic acid

A18063.06 A18063.14
A18063.06EA 2130 CZK
A18063.06 A18063.14
(2,6-Difluorophenyl)acetic acid ≥98%
(2,6-Difluorophenyl)acetic acid

Purity: 98% CAS No.: 85068-28-6 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 99-102° Beilstein Reference: 3650109

Vzorec: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
Bod varu: 259 °C (1013 hPa)
Bod tání: 99…102 °C
Hustota: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010001
CAS číslo: 85068-28-6
EINECS: 285-289-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR