(2,4-Difluorophenyl)acetic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,4-Difluorophenylacetic acid
B20431.06
B20431.14
B20431.06EA
3920
CZK
B20431.06
B20431.14
(2,4-Difluorophenyl)acetic acid ≥99%
(2,4-Difluorophenyl)acetic acid
Purity: 99%
CAS No.: 81228-09-3
Molecular Formula: C8H6F2O2
Formula Weight: 172.13
Melting Point: 117-119°
Beilstein Reference: 3649727
Vzorec:
(C₆H₃F₂)CH₂COOH MW: 172,13 g/mol Bod varu: 259 °C (1013 hPa) Bod tání: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009999 CAS číslo: 81228-09-3 EINECS: 279-709-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...