(2,4,5-Trifluorophenyl)acetic acid ≥94%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H26190.06
H26190.03
H26190.06EA
3230
CZK
H26190.06
H26190.03
(2,4,5-Trifluorophenyl)acetic acid ≥94%
(2,4,5-Trifluorophenyl)acetic acid
Purity: 94%
CAS No.: 209995-38-0
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Melting Point: 121-124°
MDL No.: MFCD00082479
Vzorec:
(C₆H₂F₃)CH₂COOH Bod tání: 121…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082479 CAS číslo: 209995-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...