Tisk…

(4-Phenoxyphenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Phenoxyphenylacetic Acid

M-Clarity™ quality level = MQ100

665053-1G 665053-5G
SIAL665053-1GEA 1450 CZK
SIAL665053-1G SIAL665053-5G
(4-Phenoxyphenyl)acetic acid, Sigma-Aldrich®
(4-Phenoxyphenyl)acetic acid
Vzorec: (C₆H₅OC₆H₅)CH₂COOH
MW: 228,25 g/mol
MDL Number: MFCD00079779
CAS číslo: 6328-74-1
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR