(3,4-Dihydroxyphenyl)acetic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid
M-Clarity™ quality level = MQ100
850217-1G
850217-5G
SIAL850217-1GEA
784
CZK
SIAL850217-1G
SIAL850217-5G
(3,4-Dihydroxyphenyl)acetic acid, Sigma-Aldrich®
(3,4-Dihydroxyphenyl)acetic acid
Vzorec:
(C₆H₃(OH)₂)CH₂COOH MW: 168,15 g/mol Bod varu: 337 °C (1013 hPa) Bod tání: 131…132 °C |
MDL Number:
MFCD00004338 CAS číslo: 102-32-9 EINECS: 203-024-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...