Tisk…

(4-Hydroxyphenyl)acetic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Hydroxyphenylacetic Acid

A15018.14 A15018.22 A15018.36
A15018.14EA 1760 CZK
A15018.14 A15018.22 A15018.36
(4-Hydroxyphenyl)acetic acid ≥99%
(4-Hydroxyphenyl)acetic acid

Purity: 99% CAS No.: 156-38-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 148-150° Beilstein Reference: 1448766

Vzorec: (C₆H₄(OH))CH₂COOH
MW: 152.15
Bod varu: 347
Bod tání: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004347
CAS číslo: 156-38-7
EINECS: 205-851-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR