Tisk…

(2-Hydroxyphenyl)acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Hydroxyphenylacetic acid

A13924.09 A13924.18
A13924.09EA 1280 CZK
A13924.09 A13924.18
(2-Hydroxyphenyl)acetic acid ≥98%
(2-Hydroxyphenyl)acetic acid

Purity: 98% CAS No.: 614-75-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 145-147° Beilstein Reference: 908000

Vzorec: (C₆H₄OH)CH₂COOH
MW: 152.15
Bod varu: 328
Bod tání: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS číslo: 614-75-5
EINECS: 210-393-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR