(3,4-Dimethoxyphenyl)acetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Homoveratric acid, 3,4-Dimethoxyphenylacetic acid
B20625.14
B20625.22
B20625.36
B20625.14EA
1040
CZK
B20625.14
B20625.22
B20625.36
(3,4-Dimethoxyphenyl)acetic acid ≥98%
(3,4-Dimethoxyphenyl)acetic acid
Purity: 98%
CAS No.: 93-40-3
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 95-98°
Beilstein Reference: 1110282
Vzorec:
(C₆H₃(OCH₃)₂)CH₂COOH MW: 196.2 Bod varu: 327 Bod tání: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004335 CAS číslo: 93-40-3 EINECS: 202-244-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...