Tisk…

(4-Methoxyphenyl)acetic acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Methoxyphenylacetic acid

A14399.06 A14399.22 A14399.36
A14399.06EA 865 CZK
A14399.06 A14399.22 A14399.36
(4-Methoxyphenyl)acetic acid ≥98%
(4-Methoxyphenyl)acetic acid

Purity: 98+% CAS No.: 104-01-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 84-88° Boiling Point: 138-140°/3mm MDL No.: MFCD00004345

Vzorec: (C₆H₄(OCH₃))CH₂COOH
MW: 166,18 g/mol
Bod varu: 138…140 °C (3 mmHg)
Bod tání: 84…88 °C
Bod vzplanutí: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS číslo: 104-01-8
EINECS: 203-166-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR