(4-Ethoxyphenyl)acetic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Ethoxyphenylacetic Acid
A10129.09
A10129.14
A10129.18
A10129.09EA
1160
CZK
A10129.09
A10129.14
A10129.18
(4-Ethoxyphenyl)acetic acid ≥98%
(4-Ethoxyphenyl)acetic acid
Purity: 98+%
CAS No.: 4919-33-9
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 87-91°
Beilstein Reference: 1103143
Vzorec:
(C₆H₄(OCH₂CH₃))CH₂COOH MW: 180,2 g/mol Bod varu: 313 °C (1013 hPa) Bod tání: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004346 CAS číslo: 4919-33-9 EINECS: 225-545-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...