Tisk…

(3-Methoxyphenyl)acetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A10951.06 A10951.14 A10951.22
A10951.06EA 948 CZK
A10951.06 A10951.14 A10951.22
(3-Methoxyphenyl)acetic acid ≥97%
(3-Methoxyphenyl)acetic acid

Purity: 97% CAS No.: 1798-09-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 67-72° Beilstein Reference: 2614004

Vzorec: (C₆H₄(OCH₃))CH₂COOH
Bod varu: 305,95 °C (1013 hPa)
Bod tání: 67…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00004334
CAS číslo: 1798-09-0
EINECS: 217-282-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR