(2-Methoxyphenyl)acetic acid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2-Methoxyphenylacetic acid
M-Clarity™ quality level = MQ100
180653-25G
180653-5G
SIAL180653-25GEA
2280
CZK
SIAL180653-25G
SIAL180653-5G
(2-Methoxyphenyl)acetic acid, Sigma-Aldrich®
(2-Methoxyphenyl)acetic acid
Vzorec:
(C₆H₄OCH₃)CH₂COOH MW: 166,18 g/mol Bod varu: 332 °C (1013 hPa) Bod tání: 120…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004321 CAS číslo: 93-25-4 EINECS: 202-231-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...