(4-Nitrophenyl)acetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Nitrophenylacetic acid
A15901.14
A15901.22
A15901.36
A15901.14EA
948
CZK
A15901.14
A15901.22
A15901.36
(4-Nitrophenyl)acetic acid ≥98%
(4-Nitrophenyl)acetic acid
Purity: 98%
CAS No.: 104-03-0
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 152-156°
Beilstein Reference: 1911801
Merck Reference: 14,6622
MDL No.: MFCD00007383
Vzorec:
(C₆H₄(NO₂))CH₂COOH MW: 181.15 Bod tání: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007383 CAS číslo: 104-03-0 EINECS: 203-168-5 Merck Index: 12,06719 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...