Tisk…

(3-Nitrophenyl)acetic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

B21540.03 B21540.06 B21540.14
B21540.03EA 872 CZK
B21540.03 B21540.06 B21540.14
(3-Nitrophenyl)acetic acid ≥99%
(3-Nitrophenyl)acetic acid

Purity: 99% CAS No.: 1877-73-2 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 117-120° Beilstein Reference: 2050088

Vzorec: (C₆H₄(NO₂))CH₂COOH
MW: 181.15
Bod tání: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007278
CAS číslo: 1877-73-2
EINECS: 217-512-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR