(2-Nitrophenyl)acetic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(2-Nitrophenyl)acetic acid
A13411.14
A13411.22
A13411.14EA
1710
CZK
A13411.14
A13411.22
(2-Nitrophenyl)acetic acid ≥99%
(2-Nitrophenyl)acetic acid
Purity: 99%
CAS No.: 3740-52-1
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Odour: Odourless
Melting Point: 139-142°
Beilstein Reference: 1959243
MDL No.: MFCD00007190
Vzorec:
(C₆H₄NO₂)CH₂COOH MW: 181,15 g/mol Bod tání: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007190 CAS číslo: 3740-52-1 EINECS: 223-128-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...